Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

432245

Sigma-Aldrich

Dichlorotetrakis(triphenylphosphine)ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]4RuCl2
CAS Number:
Molecular Weight:
1225.15
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

Cl[Ru]Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9.c%10ccc(cc%10)P(c%11ccccc%11)c%12ccccc%12

InChI

1S/4C18H15P.2ClH.Ru/c4*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h4*1-15H;2*1H;/q;;;;;;+2/p-2

InChI key

OIWNHEPSSHYXTG-UHFFFAOYSA-L

Application

Catalyst for:
  • Selective hydrogenation of maleic anhydride or succinic anhydride
  • Sythesis of portions of the natural antibiotic tetrodecamycin
  • Oxidation of hydroxamic acid
  • Hydroesterification reactions
  • Hydroboration of alkenes
  • Oxidation of sulfide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service