Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

432148

Sigma-Aldrich

Thulium(III) fluoride

anhydrous, powder, 99.99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TmF3
CAS Number:
Molecular Weight:
225.93
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

Quality Level

Assay

99.99%

form

powder

reaction suitability

core: thulium
reagent type: catalyst

SMILES string

F[Tm](F)F

InChI

1S/3FH.Tm/h3*1H;/q;;;+3/p-3

InChI key

AATUHDXSJTXIHB-UHFFFAOYSA-K

Application

  • Used for glass doping
  • Used to enhance mid-IR emission in oxyfluoride glass ceramics
Used to improve ultraviolet and visible upconversion fluorescence in fluorinated glasses.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Materials Research, 1-2, 101-101 (1994)
R Peslin et al.
Journal of applied physiology (Bethesda, Md. : 1985), 80(6), 2198-2203 (1996-06-01)
Specific airway resistance (sRaw) measured by body plethysmography without conditioning of the inspired air to BTPS exhibits a strong frequency dependence related to the fact that the warming and wetting of the gas in the airways is not instantaneous (R.

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service