Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

432113

Sigma-Aldrich

Lutetium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Lutetium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LuF3
CAS Number:
Molecular Weight:
231.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lutetium

impurities

<100 ppm water

SMILES string

F[Lu](F)F

InChI

1S/3FH.Lu/h3*1H;/q;;;+3/p-3

InChI key

VIHLFTMKXFWYAS-UHFFFAOYSA-K

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service