Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43119

Sigma-Aldrich

α,α-Diphenyl-N-methyl-L-prolinol

≥96.0% (sum of enantiomers, GC)

Synonym(s):

(S)-α,α-Diphenyl-N-methyl-2-pyrrolidinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO
CAS Number:
Molecular Weight:
267.37
Beilstein:
4749387
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (sum of enantiomers, GC)

optical activity

[α]20/D +55±3°, c = 1% in chloroform stab. with amylenes

mp

66-72 °C

SMILES string

CN1CCC[C@H]1C(O)(c2ccccc2)c3ccccc3

InChI

1S/C18H21NO/c1-19-14-8-13-17(19)18(20,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17,20H,8,13-14H2,1H3/t17-/m0/s1

InChI key

XIJAGFLYYNXCAB-KRWDZBQOSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ligand employed in the enantioselective addition of dialkylzincs to carbonyls

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Soai et al.
Journal of the American Chemical Society, 109, 7111-7111 (1987)
K. Soai et al.
Journal of Fluorine Chemistry, 59, 5-5 (1992)
Tetrahedron Asymmetry, 3, 677-677 (1992)
K. Soai et al.
Chemical Reviews, 92, 833-833 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service