Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

430897

Sigma-Aldrich

Rubidium formate

98%

Synonym(s):

Formic acid rubidium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2Rb
CAS Number:
Molecular Weight:
132.50
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

98%

form

solid

SMILES string

[Rb+].[H]C([O-])=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service