Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

430846

Sigma-Aldrich

Poly(ethylene-co-ethyl acrylate-co-maleic anhydride)

powder, melt index 5 g/10 min (190°C/2.16kg)

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H8O2 · C4H2O3 · C2H4)x
CAS Number:
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

form

powder

melt index

5 g/10 min (190°C/2.16kg)

mp

92 °C

transition temp

softening point 75 °C (Vicat)

density

0.94 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Component in protective coatings, adhesion promoter.

Features and Benefits

Contains anhydride functionality available for derivatizing or cross-linking.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service