Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

430242

Sigma-Aldrich

Neopentyl glycol diglycidyl ether, brominated

mol wt 250-300 g/epoxide by perchloric acid method, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

mol wt

250-300 g/epoxide by perchloric acid method

extent of labeling

30 wt. % Br

impurities

≤10 ppm free epichlorohydrin
8 wt. % Cl

refractive index

n20/D 1.512 (lit.)

viscosity

275-500 cSt(25 °C, Ubbelohde)(lit.)

mp

~254 °C (dec.) (lit.)

density

1.504 g/mL at 25 °C (lit.)

Application

Flame retardant additive in potting compounds, adhesives and laminating compounds.

Features and Benefits

Low viscosity halogenated diepoxide for imparting flame retardancy. Compatible with aromatic epoxide resins.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service