Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

429899

Sigma-Aldrich

2,6-Dichloro-4-(trifluoromethoxy)aniline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(OCF3)NH2
CAS Number:
Molecular Weight:
246.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

30-32 °C (lit.)

SMILES string

Nc1c(Cl)cc(OC(F)(F)F)cc1Cl

InChI

1S/C7H4Cl2F3NO/c8-4-1-3(14-7(10,11)12)2-5(9)6(4)13/h1-2H,13H2

InChI key

FKISQWQHZULEEG-UHFFFAOYSA-N

General description

2,6-Dichloro-4-(trifluoromethoxy)aniline is a nitrogen compound useful in organic synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service