Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428213

Sigma-Aldrich

Poly(ethylene-co-propylene)

average Mw ~28,700, average Mn ~9,100 (GPC vs. polyethylene stds.)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(CH3)]X(CH2CH2)Y
CAS Number:
MDL number:
UNSPSC Code:
12162002
Pricing and availability is not currently available.

mol wt

average Mn ~9,100 (GPC vs. polyethylene stds.)
average Mw ~28,700

hardness

30 dmm (Penetrometer, ASTM D 5)

viscosity

200 poise(190 °C, Brookfield Thermosel)(lit.)

transition temp

Tg −22 °C
softening point 135 °C (ring and ball, ASTM E 28)

density

0.9 g/mL at 25 °C (lit.)

SMILES string

C=C.CC=C

InChI

1S/C3H6.C2H4/c1-3-2;1-2/h3H,1H2,2H3;1-2H2

InChI key

HQQADJVZYDDRJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Base polymer in hot melt adhesives and paper-laminating, extender and viscosity modifier in caulks and sealants and waterproofing agent in wire and cable applications.
Low odor, good heat stability, low color and broad compatibility with elastomers, plastics and tackifying resins.

Physical form

Amorphous copolymer.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service