Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428094

Sigma-Aldrich

Polyethylene

pellets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002

form

pellets

melt index

35 g/10 min (190°C/2.16kg)

hardness

52 (Shore D, ASTM D 2240)

Application

Injection molding of industrial containers and trash bins.

Features and Benefits

Exhibits high rigidity and good impact strength.

Preparation Note

Copolymerized with 5-10 wt. % hexene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Schwabegger et al.
Synthetic metals, 161-66(19-20), 2058-2062 (2011-11-04)
We report on C(60) based organic field effect transistors (OFETs) that are well optimized for low voltage operation. By replacing commonly used dielectric layers by thin parylene films or by utilizing different organic materials like divinyltetramethyldisiloxane-bis(benzocyclo-butene) (BCB), low density polyethylene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service