Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

427632

Sigma-Aldrich

Tributyl borate-11B

99 atom % 11B

Synonym(s):

Tributoxyborane-11B

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3O]311B
CAS Number:
Molecular Weight:
230.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 11B

Quality Level

refractive index

n20/D 1.408 (lit.)

bp

230-235 °C (lit.)

mp

−70 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

mass shift

depleted

SMILES string

CCCCO[11B](OCCCC)OCCCC

InChI

1S/C12H27BO3/c1-4-7-10-14-13(15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3/i13+0

InChI key

LGQXXHMEBUOXRP-XFLZAFPTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chawanphat Muangnoi et al.
Pharmaceutics, 11(8) (2019-08-04)
Previously, we synthesized curcumin and a succinate ester prodrug of curcumin namely curcumin diethyl disuccinate (CurDD) in the lab scale, which yielded hundred milligrams to few grams of the compounds. CurDD was found to be more stable in a phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service