Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

426695

Sigma-Aldrich

Poly(ethylene-co-methacrylic acid) sodium salt

melt index 3.9 g/10 min (190 °C/2.16kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x[CH2C(CH3)(CO2R)]y, R = H or Na
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

melt index

3.9 g/10 min (190 °C/2.16kg)

tensile impact

1,670 kJ/m2± (23°C)(ASTM D 1822S)

hardness

60 (Shore D)

transition temp

softening point 78 °C (Vicat, ASTM D 1525-70)
Tm 81-96 °C (DTA)

density

0.94 g/mL at 25 °C

SMILES string

[Na+].C=C.CC(=C)C([O-])=O

Looking for similar products? Visit Product Comparison Guide

Application

Golf ball covers, auto rub strips, athletic shoe soles, ski boots and cosmetic packaging.

Features and Benefits

Excellent impact strength and optical properties. Foamable, abrasion and solvent resistant and exhibits adhesion to various materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service