Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

426407

Sigma-Aldrich

4-Butylbenzaldehyde diethyl acetal

technical, 90%

Synonym(s):

4-Butyl-α,α-diethoxytoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3C6H4CH(OC2H5)2
CAS Number:
Molecular Weight:
236.35
Beilstein:
5524651
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

90%

refractive index

n20/D 1.478 (lit.)

bp

250 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CCCCc1ccc(cc1)C(OCC)OCC

InChI

1S/C15H24O2/c1-4-7-8-13-9-11-14(12-10-13)15(16-5-2)17-6-3/h9-12,15H,4-8H2,1-3H3

InChI key

VZIJDIWCWDYVFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Butylbenzaldehyde diethyl acetal is a benzaldehyde dialkyl acetal. Acetals are organic compounds derived from aldehydes which contain two single-bonded oxygen atoms attached to the same carbon atom.

Application

4-Butylbenzaldehyde diethyl acetal may be used as a starting material in the synthesis of 4-butylbenzaldehyde and 4-butylbenzyl bromide.

Other Notes

Remainder ethyl 4-butylbenzoate

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francis Giraud et al.
Journal of enzyme inhibition and medicinal chemistry, 24(5), 1067-1075 (2009-06-27)
A new series of 1-benzyl-3-(imidazol-1-ylmethyl)indoles were synthesized according to a previous 3D-QSAR predictive model and assayed for their antiparasitic activity upon Leishmania mexicana promastigotes. The biological results obtained for these twelve molecules showed an IC(50) ranging from 2.3 to 32

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service