Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

425710

Sigma-Aldrich

Samarium(III) trifluoromethanesulfonate

98%

Synonym(s):

Samarium(III) triflate, Trifluoromethanesulfonic acid samarium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm(SO3CF3)3
CAS Number:
Molecular Weight:
597.57
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 425710, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

98%

reaction suitability

core: samarium
reagent type: catalyst

SMILES string

[Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Sm/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

DDCWGUIPLGMBPO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

A water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 59, 3590-3590 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service