Skip to Content
MilliporeSigma
All Photos(1)

Documents

425621

Sigma-Aldrich

Diphenylgermanium dichloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2GeCl2
CAS Number:
Molecular Weight:
297.75
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.5975 (lit.)

bp

100 °C/0.005 mmHg (lit.)

density

1.415 g/mL at 25 °C (lit.)

SMILES string

Cl[Ge](Cl)(c1ccccc1)c2ccccc2

InChI

1S/C12H10Cl2Ge/c13-15(14,11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

PFPTYRFVUHDUIN-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruno Pescara et al.
Inorganic chemistry, 58(8), 4802-4811 (2019-03-29)
Germanium (Ge) nanoparticles are gaining increasing interest due to their properties that arise in the quantum confinement regime, such as the development of the band structure with changing size. While promising materials, significant challenges still exist related to the development

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service