Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

425583

Sigma-Aldrich

p-Tolyl trifluoromethanesulfonate

97%

Synonym(s):

p-Tolyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3C6H4CH3
CAS Number:
Molecular Weight:
240.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.441 (lit.)

bp

115 °C/50 mmHg (lit.)

density

1.342 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(OS(=O)(=O)C(F)(F)F)cc1

InChI

1S/C8H7F3O3S/c1-6-2-4-7(5-3-6)14-15(12,13)8(9,10)11/h2-5H,1H3

InChI key

XXSBXVHIYGQWDV-UHFFFAOYSA-N

General description

p-Tolyl trifluoromethanesulfonate (p-tolyl triflate) is an aryl triflate.

Application

p-Tolyl trifluoromethanesulfonate (p-tolyl triflate) may be used in the asymmetric α-arylation of ketones[1] and arylation of pyridine N-oxide.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct arylation of azine N-oxides with aryl triflates.
Schipper DJ, et al.
Tetrahedron, 65(26), 4977-4983 (2009)
Xuebin Liao et al.
Journal of the American Chemical Society, 130(1), 195-200 (2007-12-14)
The asymmetric alpha-arylation of ketones with aryl triflates is described, and the use of this electrophile with nickel and palladium catalysts containing a segphos derivative increases substantially the scope of highly enantioselective arylations of ketone enolates. The combination of aryl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service