Skip to Content
MilliporeSigma
All Photos(1)

Documents

425575

Sigma-Aldrich

2-Naphthyl trifluoromethanesulfonate

97%

Synonym(s):

2-Naphthyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3C10H7
CAS Number:
Molecular Weight:
276.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

30-32 °C (lit.)

SMILES string

FC(F)(F)S(=O)(=O)Oc1ccc2ccccc2c1

InChI

1S/C11H7F3O3S/c12-11(13,14)18(15,16)17-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H

InChI key

MDWRQYBWVTXIIJ-UHFFFAOYSA-N

General description

2-Naphthyl trifluoromethanesulfonate (2-naphthyl triflate) is an aryl triflate. Its ruthenium-catalyzed conversion to 2-bromonaphthalene has been reported.

Application

2-Naphthyl trifluoromethanesulfonate (2-naphthyl triflate) may be used as an arylating agent in the arylation of 1-(methoxycarbonyl)-2,5-dihydropyrrole by Heck reaction. It may be used as a substrate for coupling with Reformatsky reagent BrZnCH(CH3)(COOtC4H9) in the presence of “reduced” dichlorobis(1,1-diphenylphosphino)ferrocene catalyst.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of the catalyst on the Pd0-mediated reactions of BrZnCH(CH3) COOtBu with vinyl and aryl triflates.
Orsini F, et al.
Journal of Organometallic Chemistry, 367(3), 375-382 (1989)
Regiochemical control and suppression of double bond isomerization in the Heck arylation of 1-(methoxycarbonyl)-2, 5-dihydropyrrole.
Sonesson C, et al.
The Journal of Organic Chemistry, 61(14), 4756-4763 (1996)
Xiaoqiang Shen et al.
Journal of the American Chemical Society, 132(40), 14076-14078 (2010-09-23)
The palladium-catalyzed conversion of aryl and vinyl triflates to aryl and vinyl halides (bromides and chlorides) has been developed using dialkylbiaryl phosphine ligands. A variety of aryl, heteroaryl, and vinyl halides can be prepared via this method in good to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service