Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

425559

Sigma-Aldrich

Chlorotriphenyllead(IV)

technical grade, 90%

Synonym(s):

Triphenyllead(IV) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3PbCl
CAS Number:
Molecular Weight:
473.96
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical grade

Assay

90%

mp

207 °C (lit.)

SMILES string

Cl[Pb](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.ClH.Pb/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H;/q;;;;+1/p-1

InChI key

RJSSWRDMHBJWKN-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kleszczyńska et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 56(9-10), 853-856 (2001-11-29)
The influence of some inorganic (K+, Mg2+, and Al3+) and organic CnH(2n+1)SO3-, n = 12 and 14) ions was studied on the hemolysis of erythrocytes (RBC) caused by organolead compounds (tripropyllead--TPL, tributyllead--TBL and triphenyllead--TPhL chlorides). It was found that sulfonate
H Kleszczyńska et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 55(9-10), 764-769 (2000-12-01)
The physiological and hemolytic toxicities of triphenyllead chloride (TPhL) as well as its modifying influence on model lipid membranes were studied. The experiments allowed the determination of TPhL concentrations causing 50% inhibition of growth of Spirodela oligorrhiza, Lemna minor and
Janina Gabrielska et al.
Ecotoxicology and environmental safety, 89, 36-42 (2012-12-25)
Previously we have shown a toxic effect of the organometallic compound triphenyllead (TPhPb) on cells. In the present study we evaluated the destructive effect of TPhPb on model systems--serum albumin and liposome membranes--alone and under UVB irradiation. UVB irradiation of
Antoni Murkowski et al.
Bulletin of environmental contamination and toxicology, 84(2), 157-160 (2009-09-29)
The effect of the organometallic compounds containing lead, (C6H5)3PbCl , and tin, (C6H5)3SnCl, on Chlorella green algae photosystem II was studied. Suspension of the algae treated with (C6H5)3SnCl at concentrations of 1.0 and 4.0 micromol dm(-3) for 22 h revealed
Dariusz Man et al.
Journal of biomedicine & biotechnology, 2009, 969480-969480 (2009-07-17)
Biophysical activity of two twin organometallic compounds Triphenyltin chloride (TPhT) and Triphenyllead chloride (TPhL) in their interreaction with model membranes, as well as with yeast cells Saccharomyces cerevisiae, was investigated. Four measurement methods were used in the experiments: two physical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service