Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

425095

Sigma-Aldrich

1-(Trimethylsilyl)-1H-benzotriazole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3Si
CAS Number:
Molecular Weight:
191.31
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

96%

form

liquid

refractive index

n20/D 1.545 (lit.)

bp

125-126 °C/3 mmHg (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)n1nnc2ccccc12

InChI

1S/C9H13N3Si/c1-13(2,3)12-9-7-5-4-6-8(9)10-11-12/h4-7H,1-3H3

InChI key

VHOLOZYRMQVHBK-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

26.6 °F - closed cup

Flash Point(C)

-3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service