Skip to Content
MilliporeSigma
All Photos(1)

Documents

424919

Sigma-Aldrich

4-Benzyloxy-3-methoxyphenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H3(OCH3)CH2CO2H
CAS Number:
Molecular Weight:
272.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

100-102 °C (lit.)

SMILES string

COc1cc(CC(O)=O)ccc1OCc2ccccc2

InChI

1S/C16H16O4/c1-19-15-9-13(10-16(17)18)7-8-14(15)20-11-12-5-3-2-4-6-12/h2-9H,10-11H2,1H3,(H,17,18)

InChI key

AYCIHUSEBJLTBF-UHFFFAOYSA-N

General description

4-Benzyloxy-3-methoxyphenylacetic acid is an aromatic phenyl acetic acid. It has been synthesized by the benzylation of 4-hydroxy-3-methoxyphenylacetic acid.

Application

4-Benzyloxy-3-methoxyphenylacetic acid may be used as one of the reactant in the synthesis of 1,2-bis(4-benzyloxy-3-methoxyphenyl)-3-hydroxy-propionic acid.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of the erythro and threo Forms of 1,2-Bis(4-hydroxy-3-methoxyphenyl)-1,3-propanediol.
Berndtsson I, et al.
Acta Chemica Scandinavica. Series B, 34(6), 453-455 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service