Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

424870

Sigma-Aldrich

4-Bromo-3-nitrotoluene

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Br
CAS Number:
Molecular Weight:
216.03
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

bp

151.5-152.5 °C/14 mmHg (lit.)

mp

31-33 °C (lit.)

density

1.578 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Br)c(c1)[N+]([O-])=O

InChI

1S/C7H6BrNO2/c1-5-2-3-6(8)7(4-5)9(10)11/h2-4H,1H3

InChI key

UPBUTKQMDPHQAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromo-3-nitrotoluene (2-bromo-5-methylnitrobenzene, 1-bromo-4-methyl-2-nitrobenzene) is a halonitrobenzene. It has been synthesized from 4-methyl-2-nitroaniline and confirmed by 1H NMR. The crystal structure of 1-bromo-4-methyl-2-nitrobenzene is found to be orthorhombic in the Pna21 space group. Its enthalpy of vaporization at boiling point has been determined.

Application

4-Bromo-3-nitrotoluene may be used as a starting material in the synthesis of 2-bromo-5-methylaniline and as a reagent in the synthesis of 2-nitro-4:4′-dimethyl-diphenyl by reacting with p-iodotoluene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biaryl Product Formation from Cross-coupling in Palladiumcatalyzed Borylation of a Boc Protected Aminobromoquinoline Compound.
Fang H, et al.
Molecules (Basel), 9(3), 178-184 (2004)
1-Bromo-4-methyl-2-nitrobenzene.
Li P, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(10), 2641-2641 (2011)
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 537-537 (2014)
Orientation effects in the diphenyl series. Part XII. The mono-and the di-nitration of 4: 4'-dimethyldiphenyl.
Marler EEJ and Turner EE.
Journal of the Chemical Society, 2391-2394 (1932)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service