Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

423971

Sigma-Aldrich

Ethyl 4-pyridylacetate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.499 (lit.)

bp

127-128 °C/14 mmHg (lit.)

mp

18-19 °C (lit.)

density

1.079 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)Cc1ccncc1

InChI

1S/C9H11NO2/c1-2-12-9(11)7-8-3-5-10-6-4-8/h3-6H,2,7H2,1H3

InChI key

QVLJLWHOILVHJJ-UHFFFAOYSA-N

General description

Ethyl 4-pyridylacetate is an ethyl ester of 4-pyridyl acetic acid. Its hydrochloride salt was used to prepare the starting material required for the synthesis of 2,2-dideutero-1-azabicyclo(2,2,2)-octane. It participates as a reagent in the synthesis of triarylethane phosphodiesterase 4 inhibitors.

Application

Ethyl 4-pyridylacetate is a pyridine-based compound used for the grafting onto the self-adhesive gold substrates.
It may be used for the the following:
  • As a starting material in the synthesis of 4-piperidylethanol.
  • As a reactant in the synthesis of 4-(pyridyl) isosteres of meperidine.
  • As a starting material in the synthesis of ethyl-4-piperidylacetate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antinociceptive activity of 2-and 3-methyl derivatives of 4-(pyridyl) isosteres of meperidine.
Buolamwini JK and Knaus EE.
Drug Des. Discovery, 8(2), 145-156 (1991)
The Pyridylthioacetmorpholides and Pyridylacetic Acids.
Malan RL and Dean PM.
Journal of the American Chemical Society, 69(7), 1797-1798 (1947)
Covalent grafting onto self-adhesive surfaces based on aryldiazonium salt seed layers.
Viel P, et al.
Journal of Materials Chemistry, 18(48), 5913-5920 (2008)
CDP840. A prototype of a novel class of orally active anti-inflammatory phosphodiesterase 4 inhibitors.
Alexander RP, et al.
Bioorganic & Medicinal Chemistry Letters, 12(11), 1451-1456 (2002)
Azabicyclo compounds. X. The preparation of 1-azabicyclo (2, 2, 2) octanes deuterated at the positions 2, 3 and 4.
Palecek, J.
Collection of Czechoslovak Chemical Communications, 36(5), 2070-2074 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service