Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422940

Sigma-Aldrich

4,6-Dimethyl-2-nitroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H2(NO2)NH2
CAS Number:
Molecular Weight:
166.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

64-68 °C (lit.)

SMILES string

Cc1cc(C)c(N)c(c1)[N+]([O-])=O

InChI

1S/C8H10N2O2/c1-5-3-6(2)8(9)7(4-5)10(11)12/h3-4H,9H2,1-2H3

InChI key

VSRYYONYIUUFFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,6-Dimethyl-2-nitroaniline (2,4-Dimethyl-6-nitroaniline), an aromatic amine, is an important aromatic organic intermediate. It has been synthesized from 2,4-dimethylaniline and the crystal structure has been investigated. It has been reported to be determined by diazotization and coupling spectrophotometric technique using 8-amino-1-hydroxynaphthalene-3,6-disulphonic acid and N-(1-naphthyl)ethylenediamine as coupling agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Norwitz et al.
Talanta, 33(4), 311-314 (1986-04-01)
Twenty more aromatic amines are determined by the diazotization and coupling spectrophotometric technique, with 8-amino-1-hydroxynaphthalene-3,6-disulphonic acid (H-acid) and N-(1-naphthy)ethylenediamine (N-na) as coupling agents. The following are determined by both methods: 2,4-diaminotoluene, 2-aminobenzotrifluoride, 4-benzoxyaniline, 2,4-dimethyl-6-nitroaniline, 4,5-dimethyl-2-nitroaniline, 2-amino-9-fluorenone, naphthionic acid (sodium salt)
2, 4-Dimethyl-6-nitroaniline.
Chen HK.
Acta Crystallographica Section E, Structure Reports Online, 68(5), 1392-1392 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service