Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422711

Sigma-Aldrich

Acetone O-(benzyloxycarbonyl)oxime

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=NOCO2CH2C6H5
CAS Number:
Molecular Weight:
207.23
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

46-48 °C (lit.)

SMILES string

C\C(C)=N\OC(=O)OCc1ccccc1

InChI

1S/C11H13NO3/c1-9(2)12-15-11(13)14-8-10-6-4-3-5-7-10/h3-7H,8H2,1-2H3

InChI key

YAECLTQWHQOIQR-UHFFFAOYSA-N

General description

Acetone O-(benzyloxycarbonyl)oxime is an oxime derivative. It is an organic building block used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mao Zhang et al.
Frontiers in cardiovascular medicine, 8, 820416-820416 (2022-02-08)
Heart failure is a syndrome in which the heart cannot pump enough blood to meet the body's needs, resulting from impaired ventricular filling or ejection of blood. Heart failure is still a global public health problem and remains a substantial
P Burkert et al.
The European journal of neuroscience, 23(2), 335-349 (2006-01-20)
Insect metamorphosis is a compelling example of postembryonic remodelling of neuronal structure and synaptic connectivity as larval and adult behaviours place distinct demands on the CNS. Holometabolous insects such as the moth Manduca sexta have long served as suitable models

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service