Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422592

Sigma-Aldrich

Dilithium tetrabromonickelate(II) solution

0.5 M in THF

Synonym(s):

Lithium tetrabromonickelate(II) solution, di-Lithium tetrabromonickelate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2NiBr4
CAS Number:
Molecular Weight:
392.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

reaction suitability

core: nickel
reagent type: catalyst

concentration

0.5 M in THF

bp

67 °C

density

1.065 g/mL at 25 °C

SMILES string

[Li+].[Li+].Br[Ni--](Br)(Br)Br

InChI

1S/4BrH.2Li.Ni/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

AVJDNPHXGKKFIA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for the regioselective preparation of magnetic resonance contrast agents

Reagent for:
  • Regioselective synthesis of (-)-gallocatechin
  • Synthesis of ATP mimics
  • Reaction with unsaturated epoxides

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service