Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422568

Sigma-Aldrich

2-Deoxy-2,2-difluoro-D-erythro-pentonic acid γ-lactone 3,5-dibenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14F2O6
CAS Number:
Molecular Weight:
376.31
MDL number:
UNSPSC Code:
12352103

optical activity

[α]25/D +62°, c = 1 in ethyl acetate

mp

117-119 °C (lit.)

SMILES string

FC1(F)[C@H](OC(=O)c2ccccc2)[C@@H](COC(=O)c3ccccc3)OC1=O

Application

Useful for the preparation of difluorinated pyrimidine nucleosides.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 565-565 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service