Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422517

Sigma-Aldrich

2,4,6-Trimethoxypyrimidine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O3
CAS Number:
Molecular Weight:
170.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

51-54 °C (lit.)

SMILES string

COc1cc(OC)nc(OC)n1

InChI

1S/C7H10N2O3/c1-10-5-4-6(11-2)9-7(8-5)12-3/h4H,1-3H3

InChI key

RJVAFLZWVUIBOU-UHFFFAOYSA-N

General description

2,4,6-Trimethoxypyrimidine is a pyrimidine derivative. One of the methods reported for its synthesis is by the reaction of 2,4,6-trichloropyrimidine with sodium ethoxide at 70-100°C.[1] Its transformation into 1,6-dihydro-2,4-dimethoxy-1-methyl-6-oxopyrimidine by Hilbert-Johnson reaction has been reported.

Application

2,4,6-Trimethoxypyrimidine may be used as a model compound in a study to determine the qualitative composition of mixtures formed during the methylation of barbituric acid and its derivatives by diazomethane.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvation effects in the methylation of barbituric acid and its derivatives by diazomethane.
Krasnov KA, et al.
Chemistry of Heterocyclic Compounds, 23(11), 1218-1221 (1987)
Chemistry of heterocyclic compounds. 29. Synthesis and reactions of multihetero macrocycles possessing 2, 4-pyrimidino subunits connected by carbon-oxygen and/or-sulfur linkages.
Newkome GR, et al.
The Journal of Organic Chemistry, 43(17), 3362-3367 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service