Skip to Content
MilliporeSigma
All Photos(1)

Documents

422509

Sigma-Aldrich

7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

99%

Synonym(s):

5-Methyl[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 6-Methyl-4-hydroxy-1,3,3a,7-tetraazaindene, 7-Hydroxy-5-methyl-1,3,4-triazaindolizine, NSC 32071, NSC 511493

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O
CAS Number:
Molecular Weight:
150.14
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

99%

mp

280-283 °C (lit.)

SMILES string

Cc1cc(O)n2ncnc2n1

InChI

1S/C6H6N4O/c1-4-2-5(11)10-6(9-4)7-3-8-10/h2-3,11H,1H3

InChI key

ZUIVNYGZFPOXFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for the synthesis of:
  • Ruthenium(II)-Hmtpo complexes
  • Dihydroorotate dehydrogenase inhibitors with antimalarial activity

Reactant for the Vilsmeier reaction of conjugated carbocycles and heterocycles

Investigations of the pharmacological activity caused by binding to HIV TAR RNA

Additive to study the space charge layer in silver bromide microcrystals

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service