Skip to Content
MilliporeSigma
All Photos(1)

Documents

422223

Sigma-Aldrich

3-(Trifluoroacetyl)indole

99%

Synonym(s):

3-Indolyl trifluoromethyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6F3NO
CAS Number:
Molecular Weight:
213.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

211-214 °C (lit.)

SMILES string

FC(F)(F)C(=O)c1c[nH]c2ccccc12

InChI

1S/C10H6F3NO/c11-10(12,13)9(15)7-5-14-8-4-2-1-3-6(7)8/h1-5,14H

InChI key

LCMDCXWSHDFQKP-UHFFFAOYSA-N

General description

3-(Trifluoroacetyl)indole is a heterocyclic trifluoromethyl ketone. One of the methods reported for its synthesis is by the reaction of indole with trifluoroacetic anhydride.

Application

  • Reactant for enantioselective synthesis of indoles via palladium-catalyzed kinetic asymmetrical alkylation
  • Reactant for preparation of N-pyridinyl indolecarboxamides via amidation of aminopyridine derivatives. with indolecarboxylic acid
  • Reactant for preparation of mast cell tryptase inhibitors, starting from indoles; using amidation as the key step
  • Reactant for formation of Michael adducts via Baylis-Hillman reaction
  • Reactant for preparation of indolecarboxamides via haloform cleavage of (trifluoroacetyl)indole with lithium dialkylamides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of indole-3-carboxamides via a haloform cleavage reaction of 3-trifluoroacetylindole with lithium dialkylamides.
Hassinger HL, et al
Tetrahedron Letters, 39(20), 3095-3098 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service