Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

421871

Sigma-Aldrich

(R)-(+)-α-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol

≥99%, ChiraSelect

Synonym(s):

(R)-(+)-1-(Pentafluorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6F5)OH
CAS Number:
Molecular Weight:
212.12
Beilstein:
5268492
MDL number:
UNSPSC Code:
12352103

product line

ChiraSelect

Assay

≥99%

optical activity

[α]20/D +7.5°, c = 1 in pentane

mp

41-42 °C (lit.)

SMILES string

C[C@@H](O)c1c(F)c(F)c(F)c(F)c1F

InChI

1S/C8H5F5O/c1-2(14)3-4(9)6(11)8(13)7(12)5(3)10/h2,14H,1H3/t2-/m1/s1

InChI key

WYUNHWKTLDBPLE-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service