Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

421693

Sigma-Aldrich

(S)-(−)-2-(Boc-amino)-3-cyclohexyl-1-propanol

90%

Synonym(s):

(S)-(−)-2-(tert-Butoxycarbonylamino)-3-cyclohexyl-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
257.37
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
90%

Assay

90%

form

liquid

optical activity

[α]20/D −25°, c = 1 in ethanol

refractive index

n20/D 1.464 (lit.)

bp

214 °C (lit.)

density

1.126 g/mL at 25 °C (lit.)

functional group

amine
hydroxyl

SMILES string

CC(C)(C)OC(=O)N[C@H](CO)CC1CCCCC1

InChI

1S/C14H27NO3/c1-14(2,3)18-13(17)15-12(10-16)9-11-7-5-4-6-8-11/h11-12,16H,4-10H2,1-3H3,(H,15,17)/t12-/m0/s1

InChI key

BOJQBBXPSVGTQT-LBPRGKRZSA-N

Application

Precursor to (S)-N-t-Boc-cyclohexylalaninal.[1] This precursor was oxidized to the corresponding α-amino aldehyde in good yield and high optical purity.[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.R. Leanna et al.
Tetrahedron Letters, 33, 5029-5029 (1992)
D.J. Krysan et al.
Organic preparations and procedures international, 25, 437-437 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service