Skip to Content
MilliporeSigma
All Photos(2)

Documents

420697

Sigma-Aldrich

N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine

96%

Synonym(s):

N-(Methoxymethyl)-N-(trimethylsilylmethyl)-N-benzylamine, N-Benzyl-N-(methoxymethyl)trimethylsilylmethylamine, Benzyl(methoxymethyl)[(trimethylsilyl)methyl]amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(CH2OCH3)CH2Si(CH3)3
CAS Number:
Molecular Weight:
237.41
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

76 °C/0.3 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

COCN(Cc1ccccc1)C[Si](C)(C)C

InChI

1S/C13H23NOSi/c1-15-11-14(12-16(2,3)4)10-13-8-6-5-7-9-13/h5-9H,10-12H2,1-4H3

InChI key

RPZAAFUKDPKTKP-UHFFFAOYSA-N

Application

Forms azomethine ylides which readily undergo [3+2] cycloaddition to α,ß-unsaturated esters affording N-benzyl substituted pyrrolidines in good yields.
Reacted in asymmetric 1,3-dipolar cycloadditions in the practical, large-scale synthesis of chiral pyrrolidines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kotian, Pravin L.; et al.
Organic Process Research & Development, 9(2), 193-197 (2005)
Synthesis, 2646-2646 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service