Skip to Content
MilliporeSigma
All Photos(1)

Documents

42000101

Sigma-Aldrich

PRONOVA® UP MVG

medium viscosity sodium alginate, viscosity >200 mPa.s, 1 %(20 °C)(low), guluronate monomer units ≥60 %

Synonym(s):

High Molecular Weight TM=["PRONOVA"] UP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

form

powder

mol wt

Mw >200 kDa (approximate)

composition

guluronate monomer units, ≥60%
G/M ratio, ≥1.5

impurities

≤100 CFU/g Total viable count
≤100 EU/g Endotoxins

pH

5.5-8.5 (20 °C, 1% in water)

viscosity

>200 mPa.s, 1 %(20 °C) (
low)

General description

PRONOVA UP MVG is a medium viscosity (greater than 200 mPas) sodium alginate where minimum 60% of the monomer units are guluronate.

Application

Examples of Pronova Alginates applications include: encapsulation of living cells and therapeutic proteins for cell therapy medications, cryoprotectant, bone putty binder for tissue engineering, 3D bioprinting, foams and cell tissue scaffolds, hydrogels, advanced drug delivery and implants, wound management, and medical devices.

Legal Information

Product of Dupont Nutrition USA, Inc.
PRONOVA is a registered trademark of International N&H USA, Inc.

Disclaimer

NOT FOR USE IN HUMANS

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service