Skip to Content
MilliporeSigma
All Photos(1)

Documents

419230

Sigma-Aldrich

(Benzyloxycarbonylmethyl)triphenylphosphonium bromide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2O2CCH2P(C6H5)3Br
CAS Number:
Molecular Weight:
493.37
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

mp

124 °C (dec.) (lit.)

SMILES string

[Br-].O=C(C[P+](c1ccccc1)(c2ccccc2)c3ccccc3)OCc4ccccc4

InChI

1S/C27H24O2P.BrH/c28-27(29-21-23-13-5-1-6-14-23)22-30(24-15-7-2-8-16-24,25-17-9-3-10-18-25)26-19-11-4-12-20-26;/h1-20H,21-22H2;1H/q+1;/p-1

InChI key

AVQABZVTPWROCF-UHFFFAOYSA-M

Application

Reactant for:
  • Atypical aza-Morita-Baylis-Hillman mechanism
  • Thermal decomposition
  • Double aza-Michael reaction
  • Cyclodextrin derivatives
  • Stereoselective preparation of α,β-unsaturated carbonyl compounds via stereoselective Wittig olefination with aldehydes under solventless conditions or ultrasonication
  • Synthesis of [difluoro(alkenylphenyl)methyl]phosphonic acids on non-crosslinked polystyrene as inhibitors of PTP-1B via Wittig reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service