Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

418587

Sigma-Aldrich

Poly(melamine-co-formaldehyde) butylated, solution

average Mn ~1,250, 95 wt. % in 1-butanol

Synonym(s):

1,3,5-Triazine-2,4,6-triamine, polymer with formaldehyde, butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

liquid

mol wt

average Mn ~1,250

concentration

95 wt. % in 1-butanol

refractive index

n20/D 1.49

density

1.03 g/mL at 25 °C

InChI

1S/C3H6N6.CH2O/c4-1-7-2(5)9-3(6)8-1;1-2/h(H6,4,5,6,7,8,9);1H2

InChI key

IVJISJACKSSFGE-UHFFFAOYSA-N

Application

Cross-linking agent.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service