Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

418366

Sigma-Aldrich

Poly(ethylene glycol) butyl ether

average Mn ~206

Synonym(s):

α-Butyl-ω-hydroxypoly(oxyethylene), Polyethylene glycol mono(n -butyl) ether, Polyoxyethylene monobutyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

mol wt

average Mn ~206

refractive index

n20/D 1.44 (lit.)

density

0.99 g/mL at 25 °C (lit.)

Ω-end

hydroxyl

α-end

ether

SMILES string

OCCO.CCCCO

InChI

1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3

InChI key

POAOYUHQDCAZBD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service