Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417823

Sigma-Aldrich

4-Chloro-2-(1H-pyrazol-3-yl)phenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7ClN2O
CAS Number:
Molecular Weight:
194.62
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

155-158 °C (lit.)

functional group

chloro

SMILES string

Oc1ccc(Cl)cc1-c2cc[nH]n2

InChI

1S/C9H7ClN2O/c10-6-1-2-9(13)7(5-6)8-3-4-11-12-8/h1-5,13H,(H,11,12)

InChI key

DMGLUMYOLOAXJY-UHFFFAOYSA-N

General description

4-Chloro-2-(1H-pyrazol-3-yl)phenol undergoes Mannich reaction with N,N′-bis(methoxymethyl)diaza-18-crown-6 to afford NCH2N-linked bis(3-(5-chloro-2-hydroxy)pyrazol-1-ylmethyl)-substituted diazacrown ether (98% yield).[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Su et al.
The Journal of organic chemistry, 64(24), 8855-8861 (2001-10-25)
Ten new 8-hydroxyquinoline-containing diaza-18-crown-6 ligands and analogues were synthesized via a one-pot or stepwise Mannich reaction, reductive amination, or by reacting diaza-18-crown-6 with 5,7-dichloro-2-iodomethyl-8-quinolinol in the presence of N,N-diisopropylethylamine. The Mannich reaction of N,N'-bis(methoxymethyl)diaza-18-crown-6 with 4-chloro-2-(1H-pyrazol-3-yl)phenol gave the NCH(2)N-linked bis(3-(5-chloro-2-hydroxy)pyrazol-1-ylmethyl)-substituted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service