Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417777

Sigma-Aldrich

(5-Bromo-2-hydroxy-phenyl)-(1-phenyl-1H-pyrazol-4-yl)ketone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11BrN2O2
CAS Number:
Molecular Weight:
343.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

143-148 °C (lit.)

functional group

bromo
ketone

SMILES string

Oc1ccc(Br)cc1C(=O)c2cnn(c2)-c3ccccc3

InChI

1S/C16H11BrN2O2/c17-12-6-7-15(20)14(8-12)16(21)11-9-18-19(10-11)13-4-2-1-3-5-13/h1-10,20H

InChI key

MPZOVJOEZZCBKJ-UHFFFAOYSA-N

General description

(5-Bromo-2-hydroxy-phenyl)-(1-phenyl-1H-pyrazol-4-yl)ketone has been identified as one of the major compound present in the methanolic extract of Rhizophora apiculata by LC/MS analysis.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Vinod Prabhu et al.
Journal of immunotoxicology, 9(4), 341-352 (2012-07-18)
A methanolic extract of Rhizophora apiculata was evaluated for its anti-inflammatory and anti-tumor activity against B16F10 melanoma cells in BALB/c mice. The administration of R. apiculata extract was shown to inhibit the solid tumor development in mice. R. apiculata treatment

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service