Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41715

Sigma-Aldrich

Chloro-dimethyl(3,3,3-trifluoropropyl)silane

≥95.0% (GC)

Synonym(s):

Dimethyl(3,3,3-trifluoropropyl)chlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10ClF3Si
CAS Number:
Molecular Weight:
190.67
Beilstein:
1741726
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.378 (lit.)

bp

118 °C (lit.)

density

1.117 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(Cl)CCC(F)(F)F

InChI

1S/C5H10ClF3Si/c1-10(2,6)4-3-5(7,8)9/h3-4H2,1-2H3

InChI key

KBAZUXSLKGQRJF-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda Caban et al.
Rapid communications in mass spectrometry : RCM, 34(19), e8860-e8860 (2020-06-14)
Fluorinated derivatization agents allow for the identification and quantification of emerging pollutants with high sensitivity, yet details of their potential applications using electron ionization are lacking. The fluorine atom itself does not effectively participate in electron ionization. Furthermore, limited information
Magda Caban et al.
Journal of chromatography. A, 1346, 107-116 (2014-05-13)
This paper discusses a new silylating reagent - dimethyl(3,3,3-trifluoropropyl)silyldiethylamine (DIMETRIS) - for the derivatisation of non-steroidal anti-inflammatory drugs (NSAIDs) prior to GC-MS analysis. DIMETRIS reacts with seven target compounds (diclofenac, ibuprofen, ketoprofen, naproxen, flurbiprofen and paracetamol, as well as salicylic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service