Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41710

Sigma-Aldrich

N,N-Dimethyltridecylamine

≥97.0%

Synonym(s):

1-(Dimethylamino)tridecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12N(CH3)2
CAS Number:
Molecular Weight:
227.43
Beilstein:
2234660
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

refractive index

n20/D 1.439

bp

64-65 °C/0.15 mmHg (lit.)

density

0.792 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCCCCN(C)C

InChI

1S/C15H33N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16(2)3/h4-15H2,1-3H3

InChI key

ADXNPXDFKKWVGE-UHFFFAOYSA-N

General description

N,N-Dimethyltridecylamine is a tertiary amine. Its 7-oxabicyclo[2.2.l]heptane- 2,3-dicarboxylic acid salt (endothall) in combination with pyrazon has been used as major herbicide to control annual weeds in sugar beets.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weed control in sugarbeets with herbicides and cultivation.
Wicks GA and Anderson FNI.
Weed Sci., 17(4), 456-459 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service