Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417033

Sigma-Aldrich

Diethyl 4-ethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

106-109 °C (lit.)

SMILES string

CCOC(=O)C1=C(C)NC(C)=C(C1CC)C(=O)OCC

General description

Diethyl 4-ethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate is an organic building block.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of suicide substrate on the metabolism of steroids and xenobiotics and on cytochrome P-450 apoproteins.
T R Tephly et al.
Advances in experimental medicine and biology, 197, 243-251 (1986-01-01)
M A Correia et al.
Archives of biochemistry and biophysics, 294(2), 493-503 (1992-05-01)
Suicide inactivation of hepatic cytochrome P450 (P450) enzymes 2C11, 2C6, and 3A1/A2 by 3,5-dicarbethoxy-2,6-dimethyl-4-ethyl-1,4-dihydropyridine (DDEP) in intact rats results in prosthetic heme destruction, albeit by apparently distinct mechanisms. Such heme destruction is now shown to be associated with the loss
S M Kimmett et al.
Canadian journal of physiology and pharmacology, 72(4), 397-401 (1994-04-01)
Using progesterone 21-hydroxylase as a selective substrate for P450 2C6 in phenobarbital-treated male rats, and androstenedione and progesterone 6 beta-hydroxylases as well as erythromycin N-demethylase as selective markers for P450 3A1 in dexamethasone-treated female rats, we have shown that these
K Sugiyama et al.
Chemical research in toxicology, 7(5), 633-642 (1994-09-01)
The porphyrinogenic agent 3,5-dicarbethoxy-2,6-dimethyl-4-ethyl-1,4-dihydropyridine (DDEP) is known to inactivate hepatic cytochrome P450 (P450) enzymes 2C11, 2C6, and 3A1 [Correia et al. (1987) Arch. Biochem. Biophys. 258, 436-451] by different mechanisms. The inactivation of P450 2C11 and 2C6 appears to be
D S Riddick et al.
Canadian journal of physiology and pharmacology, 68(3), 370-373 (1990-03-01)
Various rat liver cytochrome P-450 (P-450) isozymes are targets for mechanism-based inactivation by 3,5-diethoxycarbonyl-2,6-dimethyl-4-ethyl-1,4- dihydropyridine (4-ethyl DDC). Unlike rat liver, which contains multiple P-450 isozymes, rabbit lung contains only three major isozymes referred to as forms 2, 5, and 6.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service