Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

416681

Sigma-Aldrich

Bis(cyclopentadienyl)titanium(IV) bis(trifluoromethanesulfonate)

98%

Synonym(s):

Bis(cyclopentadienyl)di(triflato)titanium, Di(cyclopentadienyl)titanium(IV) bis(trifluoromethanesulfonate), Titanocene bis(trifluoromethanesulfonate), Titanocene ditriflate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10F6O6S2Ti
CAS Number:
Molecular Weight:
476.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: titanium
reagent type: catalyst

SMILES string

[Ti++].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/2C5H5.2CHF3O3S.Ti/c2*1-2-4-5-3-1;2*2-1(3,4)8(5,6)7;/h2*1-5H;2*(H,5,6,7);/q;;;;+2/p-2

InChI key

ARUXRBUQJOYABI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Transesterification
  • Stereoselective carbonyl-ene cyclization reactions
  • Sakurai reactions
  • Mukaiyama cross-aldol reactions

Reactant for synthesis of heterobimetallic bridging dinitrogen complexes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service