Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

415944

Sigma-Aldrich

2-Amino-4′-methoxyacetophenone hydrochloride

90%, technical

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4C(O)CH2NH2 · HCl
CAS Number:
Molecular Weight:
201.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

90%

form

powder

mp

190-193 °C (lit.)

functional group

amine
ketone

SMILES string

Cl.COc1ccc(cc1)C(=O)CN

InChI

1S/C9H11NO2.ClH/c1-12-8-4-2-7(3-5-8)9(11)6-10;/h2-5H,6,10H2,1H3;1H

InChI key

FZVYWBMMOSHMRS-UHFFFAOYSA-N

General description

2-Amino-4′-methoxyacetophenone hydrochloride is an organic building block.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meredith C Henderson et al.
Molecular cancer therapeutics, 8(1), 36-44 (2009-01-14)
Pancreatic carcinoma is the fourth leading cause of death from cancer. Novel targets and therapeutic options are needed to aid in the treatment of pancreatic cancer. The compound UA62784 is a novel fluorenone with inhibitory activity against the centromere protein
Arthur Y Shaw et al.
The Journal of pharmacology and experimental therapeutics, 331(2), 636-647 (2009-08-07)
A series of diaryl- and fluorenone-based analogs of the lead compound UA-62784 [4-(5-(4-methoxyphenyl)oxazol-2-yl)-9H-fluoren-9-one] was synthesized with the intention of improving upon the selective cytotoxicity of UA-62784 against human pancreatic cancer cell lines with a deletion of the tumor suppressor gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service