Skip to Content
MilliporeSigma
All Photos(1)

Documents

415545

Sigma-Aldrich

D-Galactose-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

99 atom % 13C

Quality Level

Assay

≥99% (HPLC)

form

powder

optical activity

[α]25/D +79.0, c = 2 in H2O (trace NH4OH)

mp

169-170 °C (lit.)

mass shift

M+1

SMILES string

OC[C@H]1O[13CH](O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4+,5-,6?/m1/s1/i6+1

InChI key

WQZGKKKJIJFFOK-ZGYLHYIGSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

D-Galactose-1-13C is an isotope analogue of D-galactose.

Application

D-Galactose-1-13C is used to study galactose metabolism.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C Ning et al.
Pediatric research, 48(2), 211-217 (2000-08-06)
The ability of mice deficient in galactose-1-phosphate uridyltransferase (GALT) to metabolize galactose was determined in animals weaned to a mouse chow diet for a 4-wk period. When given [14C]galactose intraperitoneally, these animals slowly oxidized the sugar, excreting only 5.5% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service