Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414956

Sigma-Aldrich

Barium calcium tungsten oxide

99.9% trace metals basis

Synonym(s):

Barium calcium tungstate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba2CaWO6
CAS Number:
Molecular Weight:
594.57
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

Assay

99.9% trace metals basis

form

powder

application(s)

battery manufacturing

SMILES string

O=[Ca].O=[Ba].O=[Ba].O=[W](=O)=O

InChI

1S/2Ba.Ca.6O.W

InChI key

JLSWGWVIBBXYRW-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service