Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414271

Sigma-Aldrich

(+)-DIP-Bromide

95%

Synonym(s):

(1S)-(+)-B-Bromodiisopinocampheylborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34BBr
CAS Number:
Molecular Weight:
365.20
UNSPSC Code:
12352101
PubChem Substance ID:

Assay

95%

form

solid

mp

62-72 °C (lit.)

SMILES string

CC1[C@H](C[C@H]2C[C@@H]1C2(C)C)B(Br)[C@H]3C[C@H]4C[C@@H]([C@@H]3C)C4(C)C

InChI

1S/C20H34BBr/c1-11-15-7-13(19(15,3)4)9-17(11)21(22)18-10-14-8-16(12(18)2)20(14,5)6/h11-18H,7-10H2,1-6H3/t11-,12?,13+,14+,15-,16-,17-,18-/m0/s1

InChI key

FKBAVEASVZAXFW-XRSIQOTJSA-N

Legal Information

DIP-Bromide is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service