Skip to Content
MilliporeSigma
All Photos(1)

Documents

413186

Sigma-Aldrich

2-Dodecyl-N-(1,2,2,6,6-pentamethyl-4-piperidinyl)succinimide

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H48N2O2
CAS Number:
Molecular Weight:
420.67
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical grade

refractive index

n20/D 1.488 (lit.)

bp

220 °C/0.7 mmHg (lit.)

density

0.963 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCC1CC(=O)N(C2CC(C)(C)N(C)C(C)(C)C2)C1=O

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service