Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

413089

Sigma-Aldrich

Gallium(II) chloride

anhydrous, 99.999% trace metals basis

Synonym(s):

Digallium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga2Cl4
CAS Number:
Molecular Weight:
281.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
anhydrous
form:
crystalline

grade

anhydrous

Assay

99.999% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

Cl[Ga]Cl.Cl[Ga]Cl

InChI

1S/4ClH.2Ga/h4*1H;;/q;;;;2*+2/p-4

InChI key

PWAPABRPCBSKKS-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Applied in the preparation of a novel gallium(I) species terminally coordinated to ruthenium.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

George N. Harakas et al.
Inorganic chemistry, 36(13), 2704-2707 (1997-06-18)
The reaction of Ru(3)(CO)(12) with Ga(2)Cl(4) in the presence of gallium metal in refluxing toluene, followed by crystallization from tetrahydrofuran, produces Ru{GaCl(THF)(2)}{GaCl(2)(THF)}(2)(CO)(3).0.5 THF (1) and Ru(2){GaCl(2)(THF)}(2)(CO)(8) (2) (THF = tetrahydrofuran). The ruthenium atom in 1 has an approximately octahedral coordination

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service