Skip to Content
MilliporeSigma
All Photos(1)

Documents

412953

Sigma-Aldrich

Methyl nonadecafluorodecanoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)8CO2CH3
CAS Number:
Molecular Weight:
528.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.308 (lit.)

bp

193 °C (lit.)

density

1.762 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C11H3F19O2/c1-32-2(31)3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)9(24,25)10(26,27)11(28,29)30/h1H3

InChI key

QJFHNYDPNSFJMR-UHFFFAOYSA-N

General description

Methyl nonadecafluorodecanoate (Methyl Perfluorodecanoate) is a fluorinated organic compound.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregg T Tomy et al.
Environmental science & technology, 38(24), 6475-6481 (2005-01-27)
An eastern Arctic marine food web was analyzed for perfluorooctanesulfonate (PFOS, C8F17SO3-), perfluorooctanoate (PFOA, C7F15COO-), perfluorooctane sulfonamide (PFOSA, C8F17SO2NH2), and N-ethylperfluorooctane sulfonamide (N-EtPFOSA, C8F17SO2NHCH2CH3) to examine the extent of bioaccumulation. PFOS was detected in all species analyzed, and mean concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service