Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

412856

Sigma-Aldrich

Diglycidyl 1,2-cyclohexanedicarboxylate

Synonym(s):

Diglycidyl hexahydrophthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O6
CAS Number:
Molecular Weight:
284.31
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.487 (lit.)

bp

200-205 °C/2 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

O=C(OCC1CO1)C2CCCCC2C(=O)OCC3CO3

InChI

1S/C14H20O6/c15-13(19-7-9-5-17-9)11-3-1-2-4-12(11)14(16)20-8-10-6-18-10/h9-12H,1-8H2

InChI key

XFUOBHWPTSIEOV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S English et al.
Contact dermatitis, 15(2), 66-68 (1986-08-01)
Allergic contact dermatitis is reported in 5 machine operatives working with a cutting oil containing an epoxy compound used as a stabilizer. Positive patch test reactions to this compound were obtained in all 5 men and were negative in 25

Questions

  1. EC Number:226-826-3の全塩素含有量はどれくらいですか?

    1 answer
    1. The total chlorine content of this product is not determined. The exact composition is considered proprietary.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service